missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)thiourea 98.0+%, TCI America™

$167.76 - $672.92

Chemical Identifiers

CAS 2646-30-2
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.11
MDL Number MFCD00041176
InChI Key MRVQULNOKCOGHC-UHFFFAOYSA-N
Synonym 1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
PubChem CID 2735620
ChEBI CHEBI:34391
IUPAC Name (4-bromophenyl)thiourea
SMILES NC(=S)NC1=CC=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44841G
View Documents
TCI America
B44841G
1 g N/A
Only null left
 
B44845G
View Documents
TCI America
B44845G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2646-30-2
231.11
MRVQULNOKCOGHC-UHFFFAOYSA-N
2735620
(4-bromophenyl)thiourea
C7H7BrN2S
MFCD00041176
1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
CHEBI:34391
NC(=S)NC1=CC=C(Br)C=C1

Specifications

2646-30-2
C7H7BrN2S
1 g
1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
NC(=S)NC1=CC=C(Br)C=C1
231.11
CHEBI:34391
≥98.0% (HPLC,N)
(4-Bromophenyl)thiourea
176°C
MFCD00041176
2811
MRVQULNOKCOGHC-UHFFFAOYSA-N
(4-bromophenyl)thiourea
2735620
231.11
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YS3500000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'