missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)thiourea 98.0+%, TCI America™

Catalog No. B44845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44845G 5 g
B44841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44845G Supplier TCI America Supplier No. B44845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2646-30-2
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.11
MDL Number MFCD00041176
InChI Key MRVQULNOKCOGHC-UHFFFAOYSA-N
Synonym 1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
PubChem CID 2735620
ChEBI CHEBI:34391
IUPAC Name (4-bromophenyl)thiourea
SMILES NC(=S)NC1=CC=C(Br)C=C1

Specifications

Melting Point 176°C
Quantity 5 g
UN Number 2811
Formula Weight 231.11
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromophenyl)thiourea
RTECSNumber YS3500000
TSCA No