missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. B5015200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5015200MG 200 mg
B50151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5015200MG Supplier TCI America Supplier No. B5015200MG

Chemical Identifiers

CAS 16839-13-7
Molecular Formula C6H6BrO3P
Molecular Weight (g/mol) 236.989
MDL Number MFCD00159409
InChI Key XDGIQCFWQNHSMV-UHFFFAOYSA-N
PubChem CID 95011
IUPAC Name (4-bromophenyl)phosphonic acid
SMILES C1=CC(=CC=C1P(=O)(O)O)Br

Specifications

Melting Point 202°C
Quantity 200 mg
Formula Weight 236.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromophenyl)phosphonic Acid
EINECSNumber (3)-2524
RTECSNumber SZ6870300
TSCA No