missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl methyl sulfone, 98+%

Catalog No. AAA1142709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1142709 10 g
AAA1142718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1142709 Supplier Thermo Scientific Chemicals Supplier No. A1142709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3466-32-8 | C7H7BrO2S | 235.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3466-32-8
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD00025065
InChI Key FJLFSYRGFJDJMQ-UHFFFAOYSA-N
Synonym 4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
PubChem CID 77014
IUPAC Name 1-bromo-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 101°C to 102°C
Quantity 10 g
Beilstein 2045675
Formula Weight 235.1
Percent Purity ≥98%
Chemical Name or Material 4-Bromophenyl methyl sulfone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-421-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only