missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl Isocyanate 98.0+%, TCI America™

Catalog No. B06365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06365G 5 g
B063625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B06365G Supplier TCI America Supplier No. B06365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2493-02-9
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002022
InChI Key CZQIJQFTRGDODI-UHFFFAOYSA-N
Synonym 4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw
PubChem CID 75609
IUPAC Name 1-bromo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Br

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 158°C
Quantity 5 g
UN Number 2206
Formula Weight 198.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenyl Isocyanate
RTECSNumber CY9035500
TSCA Yes
Recommended Storage Refrigerator