missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl isocyanate, 99%

Catalog No. p-7045463
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1083406 5 g
AAL1083414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1083406 Supplier Thermo Scientific Chemicals Supplier No. L1083406
Only null left

CAS: 2493-02-9 | C7H4BrNO | 198.02 g/mol

4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

Solubility
Sparingly soluble in water (0.40 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture.

Chemical Identifiers

CAS 2493-02-9
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002022
InChI Key CZQIJQFTRGDODI-UHFFFAOYSA-N
Synonym 4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw
PubChem CID 75609
IUPAC Name 1-bromo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Br

Specifications

Melting Point 39°C to 42°C
Boiling Point 157°C to 158°C (14 mmHg)
Flash Point 109°C (228°F)
Odor Pungent
Quantity 5 g
UN Number UN2811
Beilstein 508203
Sensitivity Moisture sensitive
Merck Index 14,1430
Solubility Information Sparingly soluble in water (0.40g/L at 25°C).
Formula Weight 198.03
Percent Purity 99%
Chemical Name or Material 4-Bromophenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-662-9
RTECSNumber CY9035500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only