missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl chloroformate, 95+%

Catalog No. AC430150010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430150010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430150010 Supplier Thermo Scientific Chemicals Supplier No. 430150010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7693-44-9 | C7H4BrClO2 | 235.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 7693-44-9
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD00013256
InChI Key IKMNJYGTSSQNSE-UHFFFAOYSA-N
Synonym 4-bromophenyl chloroformate, acmc-1bh45, 4-bromophenylcarbonochloridate, 4-bromophenyl carbonochloridate, carbonochloridic acid, 4-bromophenyl ester, 4-bromophenyl carbonochloridate;4-bromophenyl carbonochloridate
PubChem CID 5237736
IUPAC Name (4-bromophenyl) carbonochloridate
SMILES C1=CC(=CC=C1OC(=O)Cl)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.6480g/mL
Boiling Point 118°C (16.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.648
Formula Weight 235.46
Percent Purity 95+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Bromophenyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only