missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenoxyacetic Acid 98.0+%, TCI America™
$42.87 - $121.16
Chemical Identifiers
| CAS | 1878-91-7 |
|---|---|
| Molecular Formula | C8H7BrO3 |
| Molecular Weight (g/mol) | 231.045 |
| InChI Key | SZEBGAQWWSUOHT-UHFFFAOYSA-N |
| Synonym | 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy |
| PubChem CID | 74657 |
| IUPAC Name | 2-(4-bromophenoxy)acetic acid |
| SMILES | C1=CC(=CC=C1OCC(=O)O)Br |
Chemical Identifiers
| 1878-91-7 | |
| 231.045 | |
| 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy | |
| 2-(4-bromophenoxy)acetic acid |
| C8H7BrO3 | |
| SZEBGAQWWSUOHT-UHFFFAOYSA-N | |
| 74657 | |
| C1=CC(=CC=C1OCC(=O)O)Br |
Specifications
| 1878-91-7 | |
| White-Yellow | |
| 5 g | |
| SZEBGAQWWSUOHT-UHFFFAOYSA-N | |
| 2-(4-bromophenoxy)acetic acid | |
| 74657 | |
| ≥98.0% (T) | |
| 4-Bromophenoxyacetic Acid |
| 159°C | |
| C8H7BrO3 | |
| 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy | |
| C1=CC(=CC=C1OCC(=O)O)Br | |
| 231.045 | |
| 231.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes