missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenoxy)-tert-butyldimethylsilane, 97%

Catalog No. AC427090050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427090050 5 mL Glass bottle
AC427090250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427090050 Supplier Thermo Scientific Chemicals Supplier No. 427090050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67963-68-2 | C12H19BrOSi | 287.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67963-68-2
Molecular Formula C12H19BrOSi
Molecular Weight (g/mol) 287.27
MDL Number MFCD00239382
InChI Key DLGZGLKSNRKLSM-UHFFFAOYSA-N
Synonym 4-bromophenoxy tert-butyl dimethylsilane, 4-bromophenoxy-tert-butyldimethylsilane, p-bromophenoxy t-butyl dimethylsilane, 4-bromophenoxy-tert-butyl-dimethylsilane, 1-bromo-4-tert-butyldimethylsiloxy benzene, 4-bromophenol tert-butyldimethylsilyl ether, silane, 4-bromophenoxy 1,1-dimethylethyl dimethyl, 1-bromo-4-1,1-dimethylethyl dimethylsilyl oxy benzene, acmc-1b8oo
PubChem CID 4227167
IUPAC Name (4-bromophenoxy)-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.1740g/mL
Boiling Point 137°C (25.0 mmHg)
Flash Point 12°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OSi(CH3)2C(CH3)3
Quantity 5 mL
Specific Gravity 1.174
Formula Weight 287.27
Percent Purity 97%
Chemical Name or Material (4-Bromophenoxy)-tert-butyldimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only