missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol 98.0+%, TCI America™

Catalog No. B39101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39101G 1 g
B39105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39101G Supplier TCI America Supplier No. B39101G

Chemical Identifiers

CAS 106-41-2
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002313
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
PubChem CID 7808
IUPAC Name 4-bromophenol
SMILES C1=CC(=CC=C1O)Br

Specifications

Melting Point 65°C
Boiling Point 238°C
Quantity 1 g
UN Number 3077
Formula Weight 173.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenol [for Biochemical Research]
EINECSNumber (3)-0896
RTECSNumber SJ7960000
TSCA Yes