missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol, 97%

Catalog No. AC304415000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304415000 500 g Glass bottle
AC304410050 5 g Glass bottle
AC304411000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC304415000 Supplier Thermo Scientific Chemicals Supplier No. 304415000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-41-2 | C6H5BrO | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-41-2
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002313
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
PubChem CID 7808
IUPAC Name 4-bromophenol
SMILES C1=CC(=CC=C1O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Brown-Pink
Boiling Point 235°C to 236°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OH
Quantity 500 g
Beilstein 06, 198
Merck Index 15, 1439
Solubility Information Solubility in water: 25g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform and ether
Formula Weight 173.01
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-394-4

RUO – Research Use Only