missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenethyl bromide, 96%

Catalog No. p-4258814
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439530010 1 g
AC439530100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439530010 Supplier Thermo Scientific Chemicals Supplier No. 439530010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1746-28-7 | C8H8Br2 | 263.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1746-28-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
InChI Key APTDRDYSJZQPPI-UHFFFAOYSA-N
Synonym 1-bromo-4-2-bromoethyl benzene, 4-bromophenethyl bromide, 4-bromophenethylbromide, benzene,1-bromo-4-2-bromoethyl, 4-bromo-1-2-bromoethyl benzene, pubchem16698, 4-bromobenzylmethyl bromide, acmc-209e8m, 1-bromo-4-2-bromoethyl ;benzene
PubChem CID 11010825
IUPAC Name 1-bromo-4-(2-bromoethyl)benzene
SMILES C1=CC(=CC=C1CCBr)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.7300g/mL
Boiling Point 97°C to 98.5°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.73
Formula Weight 263.96
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Bromophenethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only