missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethylbiphenyl 98.0+%, TCI America™

Catalog No. B18475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18475G 5 g
B184725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18475G Supplier TCI America Supplier No. B18475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2567-29-5
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD00017869
InChI Key HZQLUIZFUXNFHK-UHFFFAOYSA-N
Synonym 4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
PubChem CID 257716
IUPAC Name 1-(bromomethyl)-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CBr

Specifications

Melting Point 85°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 247.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethylbiphenyl