Learn More
4-(Bromomethyl)phenylboronic acid, 97%
CAS: 68162-47-0 | C7H8BBrO2 | 214.85 g/mol
$205.22 - $666.88
Chemical Identifiers
| CAS | 68162-47-0 |
|---|---|
| Molecular Formula | C7H8BBrO2 |
| Molecular Weight (g/mol) | 214.85 |
| MDL Number | MFCD01318113 |
| InChI Key | PDNOURKEZJZJNZ-UHFFFAOYSA-N |
| Synonym | 4-bromomethyl phenylboronic acid, 4-bromomethylphenylboronic acid, 4-bromomethyl phenyl boronic acid, 4-bromomethyl benzeneboronic acid, 4-boronobenzyl bromide, p-bromomethyl phenylboronic acid, 4-bromomethylphenyl boronic acid, 4-bromomethylbenzeneboronic acid, boronic acid, 4-bromomethyl phenyl |
| PubChem CID | 2735593 |
| IUPAC Name | [4-(bromomethyl)phenyl]boronic acid |
| SMILES | OB(O)C1=CC=C(CBr)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC391570010
|
Thermo Scientific Chemicals
391570010 |
1 g | Glass bottle |
N/A
|
|
||||
|
AC391570050
|
Thermo Scientific Chemicals
391570050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68162-47-0 | |
| 214.85 | |
| PDNOURKEZJZJNZ-UHFFFAOYSA-N | |
| 2735593 | |
| OB(O)C1=CC=C(CBr)C=C1 |
| C7H8BBrO2 | |
| MFCD01318113 | |
| 4-bromomethyl phenylboronic acid, 4-bromomethylphenylboronic acid, 4-bromomethyl phenyl boronic acid, 4-bromomethyl benzeneboronic acid, 4-boronobenzyl bromide, p-bromomethyl phenylboronic acid, 4-bromomethylphenyl boronic acid, 4-bromomethylbenzeneboronic acid, boronic acid, 4-bromomethyl phenyl | |
| [4-(bromomethyl)phenyl]boronic acid |
Specifications
| 68162-47-0 | |
| 100.0 | |
| White | |
| 96% min. (HPLC) | |
| C7H8BBrO2 | |
| MFCD01318113 | |
| 4-bromomethyl phenylboronic acid, 4-bromomethylphenylboronic acid, 4-bromomethyl phenyl boronic acid, 4-bromomethyl benzeneboronic acid, 4-boronobenzyl bromide, p-bromomethyl phenylboronic acid, 4-bromomethylphenyl boronic acid, 4-bromomethylbenzeneboronic acid, boronic acid, 4-bromomethyl phenyl | |
| OB(O)C1=CC=C(CBr)C=C1 | |
| 214.85 | |
| 214.85 | |
| Crystalline Powder |
| 96.0 | |
| 165.0°C to 172.0°C | |
| Authentic | |
| Glass bottle | |
| BrCH2C6H4B(OH)2 | |
| 1 g | |
| PDNOURKEZJZJNZ-UHFFFAOYSA-N | |
| [4-(bromomethyl)phenyl]boronic acid | |
| 2735593 | |
| 97% | |
| 4-(Bromomethyl)phenylboronic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only