missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)phenylacetic Acid 98.0+%, TCI America™

Catalog No. B13515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13515G 5 g
B135125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13515G Supplier TCI America Supplier No. B13515G

Chemical Identifiers

CAS 13737-36-5
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00010632
InChI Key WCOCCXZFEJGHTC-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
PubChem CID 4519056
IUPAC Name 2-[4-(bromomethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CBr

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)phenylacetic Acid