missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)biphenyl, 96%

Catalog No. p-4510686
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC368950050 5 g
AC368950250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC368950050 Supplier Thermo Scientific Chemicals Supplier No. 368950050

CAS: 2567-29-5 | C13H11Br | 247.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2567-29-5
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.13
InChI Key HZQLUIZFUXNFHK-UHFFFAOYSA-N
Synonym 4-bromomethylbiphenyl, 4-bromomethyl biphenyl, 4-bromomethyl-1,1'-biphenyl, 4-phenylbenzyl bromide, 1-bromomethyl-4-phenylbenzene, bromoethylbiphenyl, 4-biphenylmethylbromide, 4-bromomethyl-biphenyl
PubChem CID 257716
IUPAC Name 1-(bromomethyl)-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CBr

Specifications

Melting Point 83.0°C to 86.0°C
Color Beige to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4CH2Br
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 247.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Bromomethyl)biphenyl
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only