missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzoic Acid 97.0+%, TCI America™

Catalog No. p-7118706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08455G 5 g
B084525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08455G Supplier TCI America Supplier No. B08455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6232-88-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00002567
InChI Key CQQSQBRPAJSTFB-UHFFFAOYSA-N
Synonym 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid
PubChem CID 22599
IUPAC Name 4-(bromomethyl)benzoic acid
SMILES C1=CC(=CC=C1CBr)C(=O)O

Specifications

Melting Point 230°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 215.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)benzoic Acid