missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzoic acid, 97%

Catalog No. AAL0337122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0337122 100 g
AAL0337106 5 g
AAL0337114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0337122 Supplier Thermo Scientific Chemicals Supplier No. L0337122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6232-88-8 | C8H7BrO2 | 215.046 g/mol

4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

Solubility
Soluble in water.

Notes
Incompatible with bases, amines and oxidizing agents.

Chemical Identifiers

CAS 6232-88-8
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00002567
InChI Key CQQSQBRPAJSTFB-UHFFFAOYSA-N
Synonym 4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid
PubChem CID 22599
IUPAC Name 4-(bromomethyl)benzoic acid
SMILES C1=CC(=CC=C1CBr)C(=O)O

Specifications

Melting Point 223°C to 227°C
Quantity 100 g
UN Number UN3261
Beilstein 1862870
Solubility Information Soluble in water.
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 228-343-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only