missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzeneboronic acid, tech. 85%

Catalog No. AAL1995303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1995303 1 g
AAL1995306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1995303 Supplier Thermo Scientific Chemicals Supplier No. L1995303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 68162-47-0 | C7H8BBrO2 | 214.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68162-47-0
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318113
InChI Key PDNOURKEZJZJNZ-UHFFFAOYSA-N
Synonym 4-bromomethyl phenylboronic acid, 4-bromomethylphenylboronic acid, 4-bromomethyl phenyl boronic acid, 4-bromomethyl benzeneboronic acid, 4-boronobenzyl bromide, p-bromomethyl phenylboronic acid, 4-bromomethylphenyl boronic acid, 4-bromomethylbenzeneboronic acid, boronic acid, 4-bromomethyl phenyl
PubChem CID 2735593
IUPAC Name [4-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(CBr)C=C1

Specifications

Melting Point ∼145°C to 150°C
Quantity 1 g
UN Number UN3261
Beilstein 3030979
Formula Weight 214.85
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-(Bromomethyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only