missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 51359-78-5 |
|---|---|
| Molecular Formula | C8H7BrO |
| Molecular Weight (g/mol) | 199.047 |
| InChI Key | XYPVBKDHERGKJG-UHFFFAOYSA-N |
| Synonym | 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde |
| PubChem CID | 11206421 |
| IUPAC Name | 4-(bromomethyl)benzaldehyde |
| SMILES | C1=CC(=CC=C1CBr)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MO07312CB
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 51359-78-5 | |
| 199.047 | |
| 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde | |
| 4-(bromomethyl)benzaldehyde |
| C8H7BrO | |
| XYPVBKDHERGKJG-UHFFFAOYSA-N | |
| 11206421 | |
| C1=CC(=CC=C1CBr)C=O |
Specifications
| 51359-78-5 | |
| C8H7BrO | |
| XYPVBKDHERGKJG-UHFFFAOYSA-N | |
| 4-(bromomethyl)benzaldehyde | |
| 11206421 | |
| 97% |
| 97% | |
| 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde | |
| C1=CC(=CC=C1CBr)C=O | |
| 199.047 | |
| 199.05 | |
| 4-(Bromomethyl)benzaldehyde |