missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)benzaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals MO07312CB
| Quantity | 250mg |
|---|
Chemical Identifiers
| 51359-78-5 | |
| 199.047 | |
| 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde | |
| 4-(bromomethyl)benzaldehyde |
| C8H7BrO | |
| XYPVBKDHERGKJG-UHFFFAOYSA-N | |
| 11206421 | |
| C1=CC(=CC=C1CBr)C=O |
Specifications
| 4-(Bromomethyl)benzaldehyde | |
| 97% | |
| 250mg | |
| XYPVBKDHERGKJG-UHFFFAOYSA-N | |
| 4-(bromomethyl)benzaldehyde | |
| 11206421 | |
| 97% |
| 51359-78-5 | |
| C8H7BrO | |
| 4-bromomethyl benzaldehyde, 4-formylbenzyl bromide, alpha-bromo-4-formyltoluene, benzaldehyde,4-bromomethyl, benzaldehyde, 4-bromomethyl, pubchem19839, p-bromomethyl benzaldehyde, 4-bromomethy benzaldehyde, 4-bromomethyl-benzaldehyde | |
| C1=CC(=CC=C1CBr)C=O | |
| 199.047 | |
| 199.05 |