Learn More
4-Bromomethyl-6,7-dimethoxycoumarin, 95%
Fluorescent labeling reagent; Used in HPLC for the detection of carboxylic acids | CAS: 88404-25-5 | C12H11BrO4 | 299.12 g/mol
Supplier: Thermo Scientific Chemicals 04340204
Description
4-Bromomethyl-6,7-dimethoxycoumarin is used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It is also used as derivatization reagent: for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography, in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry and for determination of carboxylic acids in chromatographic detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Bromomethyl-6,7-dimethoxycoumarin | |
| 200°C to 203°C | |
| MFCD00011570 | |
| UN3261 | |
| 4-bromomethyl-6,7-dimethoxycoumarin, 4-bromomethyl-6,7-dimethoxy-2h-chromen-2-one, 4-bromomethyl-6,7-dimethoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-6,7-dimethoxy, 4-bromomethyl-6,7-dimethoxy coumarin, 4-bdmc, brdmc-4,6,7, 4-bromomethyl-6,7-dimethoxy-2-oxo-2h-benzopyran | |
| JGODLBJJCNQFII-UHFFFAOYSA-N | |
| 4-(bromomethyl)-6,7-dimethoxychromen-2-one | |
| 128870 | |
| 95% |
| 88404-25-5 | |
| C12H11BrO4 | |
| 2 g | |
| 4750487 | |
| Insoluble in water. | |
| COC1=C(C=C2C(=C1)C(=CC(=O)O2)CBr)OC | |
| 299.12 | |
| 299.12 | |
| Powder |
Chemical Identifiers
| 88404-25-5 | |
| 299.12 | |
| JGODLBJJCNQFII-UHFFFAOYSA-N | |
| 128870 | |
| COC1=C(C=C2C(=C1)C(=CC(=O)O2)CBr)OC |
| C12H11BrO4 | |
| MFCD00011570 | |
| 4-bromomethyl-6,7-dimethoxycoumarin, 4-bromomethyl-6,7-dimethoxy-2h-chromen-2-one, 4-bromomethyl-6,7-dimethoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-6,7-dimethoxy, 4-bromomethyl-6,7-dimethoxy coumarin, 4-bdmc, brdmc-4,6,7, 4-bromomethyl-6,7-dimethoxy-2-oxo-2h-benzopyran | |
| 4-(bromomethyl)-6,7-dimethoxychromen-2-one |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only