missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-6,7-dimethoxycoumarin, 95%

Catalog No. p-7058873
Change view
Click to view available options
Quantity:
0.1 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4340277 0.1 g
AA4340204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4340277 Supplier Thermo Scientific Chemicals Supplier No. 04340277
Only null left

Fluorescent labeling reagent; Used in HPLC for the detection of carboxylic acids | CAS: 88404-25-5 | C12H11BrO4 | 299.12 g/mol

4-Bromomethyl-6,7-dimethoxycoumarin is used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It is also used as derivatization reagent: for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography, in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry and for determination of carboxylic acids in chromatographic detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88404-25-5
Molecular Formula C12H11BrO4
Molecular Weight (g/mol) 299.12
MDL Number MFCD00011570
InChI Key JGODLBJJCNQFII-UHFFFAOYSA-N
Synonym 4-bromomethyl-6,7-dimethoxycoumarin, 4-bromomethyl-6,7-dimethoxy-2h-chromen-2-one, 4-bromomethyl-6,7-dimethoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-6,7-dimethoxy, 4-bromomethyl-6,7-dimethoxy coumarin, 4-bdmc, brdmc-4,6,7, 4-bromomethyl-6,7-dimethoxy-2-oxo-2h-benzopyran
PubChem CID 128870
IUPAC Name 4-(bromomethyl)-6,7-dimethoxychromen-2-one
SMILES COC1=C(C=C2C(=C1)C(=CC(=O)O2)CBr)OC

Specifications

Melting Point 200°C to 203°C
Quantity 0.1 g
UN Number UN3261
Beilstein 4750487
Solubility Information Insoluble in water.
Formula Weight 299.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Bromomethyl-6,7-dimethoxycoumarin
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only