missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Bromomethyl)-2-quinolinone 97.0+%, TCI America™
Supplier: TCI America B334525G
Specifications
| 4-(Bromomethyl)-2-quinolinone | |
| 257°C | |
| MFCD03426152 | |
| 4-bromomethyl-1,2-dihydroquinoline-2-one, 4-bromomethyl quinolin-2 1h-one, 4-bromomethyl-2 1h-quinolinone, 4-bromomethyl-1,2-dihydroquinolin-2-one, 4-bromomethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-bromomethyl, 4-bromomethyl-2-quinolinone, 2-hydroxy-4-bromo methyl quinoline, 4-bromomethyl hydroquinolin-2-one, 4-bromomethylquinolinon | |
| C1=CC=C2C(=C1)C(=CC(=O)N2)CBr | |
| 238.08 | |
| 238.08 | |
| Crystalline Powder |
| 4876-10-2 | |
| C10H8BrNO | |
| 25 g | |
| BBAHJCUCNVVEQU-UHFFFAOYSA-N | |
| 4-(bromomethyl)-1H-quinolin-2-one | |
| 268296 | |
| ≥97.0% (HPLC,N) |
Chemical Identifiers
| 4876-10-2 | |
| 238.08 | |
| BBAHJCUCNVVEQU-UHFFFAOYSA-N | |
| 268296 | |
| C1=CC=C2C(=C1)C(=CC(=O)N2)CBr |
| C10H8BrNO | |
| MFCD03426152 | |
| 4-bromomethyl-1,2-dihydroquinoline-2-one, 4-bromomethyl quinolin-2 1h-one, 4-bromomethyl-2 1h-quinolinone, 4-bromomethyl-1,2-dihydroquinolin-2-one, 4-bromomethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-bromomethyl, 4-bromomethyl-2-quinolinone, 2-hydroxy-4-bromo methyl quinoline, 4-bromomethyl hydroquinolin-2-one, 4-bromomethylquinolinon | |
| 4-(bromomethyl)-1H-quinolin-2-one |
Safety and Handling
TSCA : No