missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-2-quinolinone 97.0+%, TCI America™

Catalog No. p-7122890
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33455G 5 g
B334525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33455G Supplier TCI America Supplier No. B33455G

Chemical Identifiers

CAS 4876-10-2
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.08
MDL Number MFCD03426152
InChI Key BBAHJCUCNVVEQU-UHFFFAOYSA-N
Synonym 4-bromomethyl-1,2-dihydroquinoline-2-one, 4-bromomethyl quinolin-2 1h-one, 4-bromomethyl-2 1h-quinolinone, 4-bromomethyl-1,2-dihydroquinolin-2-one, 4-bromomethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-bromomethyl, 4-bromomethyl-2-quinolinone, 2-hydroxy-4-bromo methyl quinoline, 4-bromomethyl hydroquinolin-2-one, 4-bromomethylquinolinon
PubChem CID 268296
IUPAC Name 4-(bromomethyl)-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)CBr

Specifications

Melting Point 257°C
Quantity 5 g
Formula Weight 238.08
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)-2-quinolinone