missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. aah5982103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5982103 1 g
AAH59821MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5982103 Supplier Thermo Scientific Chemicals Supplier No. H5982103
Only null left

CAS: 1029439-49-3 | C13H17BBrFO2 | 314.989 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1029439-49-3
Molecular Formula C13H17BBrFO2
Molecular Weight (g/mol) 314.989
MDL Number MFCD18434471
InChI Key QOCYLABQSVWXGE-UHFFFAOYSA-N
Synonym 2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester
PubChem CID 53217331
IUPAC Name 2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F

Specifications

Melting Point 62°C to 64°C
Quantity 1 g
Formula Weight 314.99
Percent Purity 96%
Chemical Name or Material 4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only