missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofuran-2-carboxamide, 96%

Catalog No. AAH5235506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5235506 5 g
AAH5235503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5235506 Supplier Thermo Scientific Chemicals Supplier No. H5235506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 957345-95-8 | C5H4BrNO2 | 189.996 g/mol

Reactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 957345-95-8
Molecular Formula C5H4BrNO2
Molecular Weight (g/mol) 189.996
MDL Number MFCD12755858
InChI Key UDNSRLLRTQICFU-UHFFFAOYSA-N
Synonym 4-bromo-furan-2-carboxylic acid amide
PubChem CID 57589975
IUPAC Name 4-bromofuran-2-carboxamide
SMILES C1=C(OC=C1Br)C(=O)N

Specifications

Quantity 5 g
Formula Weight 189.99
Percent Purity 96%
Chemical Name or Material 4-Bromofuran-2-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only