accessibility menu, dialog, popup

UserName

CAS RN 957345-95-8

IUPAC Name: 4-bromofuran-2-carboxamide
Synonyms: 4-bromofuran-2-carboxamide
Molecular Weight (g/mol): 190
Molecular Formula: C5H4BrNO2
InChi Key: UDNSRLLRTQICFU-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Br)=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results