missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofluorobenzene 98.0+%, TCI America™

Catalog No. p-7118726
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
B088425G 25 g
B0884100G 100 g
B0884500G 500 g
3 options

Catalog No. B088425G

Supplier: TCI America B088425G

Only null left

Chemical Identifiers

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 1993
Formula Weight 175.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromofluorobenzene
EINECSNumber (3)-0067
RTECSNumber CY9033750
TSCA Yes