missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofluorobenzene, 99%

Catalog No. p-74747
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107010100 10 mL Glass bottle
AC107011000 100 mL Glass bottle
AC107015000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107010100 Supplier Thermo Scientific Chemicals Supplier No. 107010100

CAS: 460-00-4 | C6H4BrF | 175.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -8.0°C
Color Colorless
Density 1.5930g/mL
Boiling Point 150.0°C to 153.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5260 to 1.5280
Linear Formula BrC6H4F
Quantity 10 mL
Beilstein 05, 209
Specific Gravity 1.593
Solubility Information Solubility in water: 0.14g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,acetone,soluble in ether
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromofluorobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
EINECSNumber 207-300-2
RTECSNumber CY9033750
TSCA TSCA

RUO – Research Use Only