missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodiphenyl ether, 99%, Thermo Scientific™

Catalog No. AC106890050
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106890050 5g
AC106890250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106890050 Supplier Thermo Scientific Chemicals Supplier No. 106890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-55-3
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.11
InChI Key JDUYPUMQALQRCN-UHFFFAOYSA-N
Synonym 4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenyl
PubChem CID 7565
ChEBI CHEBI:77421
IUPAC Name 1-bromo-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

Specifications

Density 1.4230g/mL
Boiling Point 305.0°C
Flash Point >113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6081
Linear Formula BrC6H4OC6H5
Quantity 5g
Beilstein 06,I,105
Specific Gravity 1.423
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid release to the environment
EINECSNumber 202-952-4

RUO – Research Use Only