missing translation for 'onlineSavingsMsg'
Learn More

4-Bromodibenzothiophene 98.0+%, TCI America™

Catalog No. p-7123544
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44491G 1 g
B44495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44491G Supplier TCI America Supplier No. B44491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97511-05-2
Molecular Formula C12H7BrS
Molecular Weight (g/mol) 263.15
MDL Number MFCD02683746
InChI Key GJXAVNQWIVUQOD-UHFFFAOYSA-N
PubChem CID 458352
IUPAC Name 6-bromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES BrC1=CC=CC2=C1SC1=C2C=CC=C1

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 1 g
Formula Weight 263.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromodibenzothiophene