missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocrotonic Acid 98.0+%, TCI America™

Catalog No. B22981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22981G 1 g
B22985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22981G Supplier TCI America Supplier No. B22981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13991-36-1
Molecular Formula C4H5BrO2
Molecular Weight (g/mol) 164.99
MDL Number MFCD00082701
InChI Key DOTGZROJTAUYFQ-OWOJBTEDSA-N
Synonym trans-4-Bromo-2-butenoic Acid
PubChem CID 6436411
IUPAC Name (2E)-4-bromobut-2-enoic acid
SMILES OC(=O)\C=C\CBr

Specifications

Melting Point 74°C
Quantity 1 g
UN Number 3261
Formula Weight 164.99
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromocrotonic Acid
TSCA No
Recommended Storage Refrigerator