accessibility menu, dialog, popup

UserName

CAS RN 13991-36-1

IUPAC Name: (2E)-4-bromobut-2-enoic acid
Synonyms: 4-bromo-trans-crotonic acid
Molecular Weight (g/mol): 164.99
Molecular Formula: C4H5BrO2
InChi Key: DOTGZROJTAUYFQ-OWOJBTEDSA-N
SMILES: OC(=O)\C=C\CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results