missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromocarbazole 98.0+%, TCI America™
$61.51 - $177.08
Chemical Identifiers
| CAS | 3652-89-9 |
|---|---|
| Molecular Formula | C12H8BrN |
| Molecular Weight (g/mol) | 246.11 |
| MDL Number | MFCD18450175 |
| InChI Key | CBJHFGQCHKNNJY-UHFFFAOYSA-N |
| PubChem CID | 58338270 |
| IUPAC Name | 4-bromo-9H-carbazole |
| SMILES | BrC1=CC=CC2=C1C1=C(N2)C=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4752200MG
|
TCI America
B4752200MG |
200 mg |
Each for $61.51
|
|
|||||
|
B47521G
|
TCI America
B47521G |
1 g |
Each for $177.08
|
|
|||||
Chemical Identifiers
| 3652-89-9 | |
| 246.11 | |
| CBJHFGQCHKNNJY-UHFFFAOYSA-N | |
| 4-bromo-9H-carbazole |
| C12H8BrN | |
| MFCD18450175 | |
| 58338270 | |
| BrC1=CC=CC2=C1C1=C(N2)C=CC=C1 |
Specifications
| 3652-89-9 | |
| C12H8BrN | |
| 200 mg | |
| BrC1=CC=CC2=C1C1=C(N2)C=CC=C1 | |
| 246.11 | |
| 246.11 | |
| Crystalline Powder |
| 107°C | |
| MFCD18450175 | |
| CBJHFGQCHKNNJY-UHFFFAOYSA-N | |
| 4-bromo-9H-carbazole | |
| 58338270 | |
| ≥98.0% (GC) | |
| 4-Bromocarbazole |
Safety and Handling
TSCA : No