accessibility menu, dialog, popup

UserName

NHBr

CAS RN 3652-89-9

IUPAC Name: 4-bromo-9H-carbazole
Synonyms: 4-bromo-9H-carbazole
Molecular Weight (g/mol): 246.11
Molecular Formula: C12H8BrN
InChi Key: CBJHFGQCHKNNJY-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C1=C(N2)C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results