missing translation for 'onlineSavingsMsg'
Learn More

4-Bromocarbazole 98.0+%, TCI America™

Catalog No. B4752200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4752200MG 200 mg
B47521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4752200MG Supplier TCI America Supplier No. B4752200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3652-89-9
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.11
MDL Number MFCD18450175
InChI Key CBJHFGQCHKNNJY-UHFFFAOYSA-N
PubChem CID 58338270
IUPAC Name 4-bromo-9H-carbazole
SMILES BrC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Melting Point 107°C
Quantity 200 mg
Formula Weight 246.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromocarbazole