missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyronitrile 97.0+%, TCI America™

Catalog No. p-7118529
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05655G 5 g
B056525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B05655G Supplier TCI America Supplier No. B05655G
Only null left

Chemical Identifiers

CAS 5332-06-9
Molecular Formula C4H6BrN
Molecular Weight (g/mol) 148.00
MDL Number MFCD00001971
InChI Key CQPGDDAKTTWVDD-UHFFFAOYSA-N
Synonym 4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide
PubChem CID 21412
IUPAC Name 4-bromobutanenitrile
SMILES C(CC#N)CBr

Specifications

Boiling Point 205°C
Quantity 5 g
UN Number 3276
Formula Weight 148.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromobutyronitrile
RTECSNumber ET8925000
TSCA Yes