missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyronitrile, 97%

Catalog No. p-7080925
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1067409 10 g
AAA1067418 50 g
AAA1067430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1067409 Supplier Thermo Scientific Chemicals Supplier No. A1067409
Only null left

CAS: 5332-06-9 | C4H6BrN | 148.00 g/mol

4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.

Solubility
Miscible with toluene and benzene. Immiscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 5332-06-9
Molecular Formula C4H6BrN
Molecular Weight (g/mol) 148.00
MDL Number MFCD00001971
InChI Key CQPGDDAKTTWVDD-UHFFFAOYSA-N
Synonym 4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide
PubChem CID 21412
IUPAC Name 4-bromobutanenitrile
SMILES C(CC#N)CBr

Specifications

Density 1.491
Boiling Point 204°C to 206°C
Flash Point 103°C (217°F)
Odor Odorless
Refractive Index 1.478
Quantity 10 g
UN Number UN3276
Beilstein 956577
Solubility Information Miscible with toluene and benzene. Immiscible with water.
Formula Weight 148.01
Percent Purity 97%
Chemical Name or Material 4-Bromobutyronitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 226-236-6
RTECSNumber ET8925000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only