missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyronitrile, 97%

Catalog No. p-73192
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106800050 5 mL Glass bottle
AC106800250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106800050 Supplier Thermo Scientific Chemicals Supplier No. 106800050

CAS: 5332-06-9 | C4H6BrN | 148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5332-06-9
Molecular Formula C4H6BrN
Molecular Weight (g/mol) 148
MDL Number MFCD00001971
InChI Key CQPGDDAKTTWVDD-UHFFFAOYSA-N
Synonym 4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide
PubChem CID 21412
IUPAC Name 4-bromobutanenitrile
SMILES C(CC#N)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4800g/mL
Boiling Point 205.0°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.4790
Linear Formula Br(CH2)3CN
Quantity 5 mL
Beilstein 02, 283
Specific Gravity 1.48
Solubility Information Solubility in water: immiscible
Formula Weight 148
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromobutyronitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-236-6
RTECSNumber ET8925000
TSCA TSCA

RUO – Research Use Only