missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric acid, 97%

Catalog No. p-73181
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133470050 5 g Glass Bottle
AC133470250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133470050 Supplier Thermo Scientific Chemicals Supplier No. 133470050

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 28°C to 32°C
Color Brown
Boiling Point 301°C
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Br(CH2)3COOH
Quantity 5 g
Beilstein 02, 283
Solubility Information Solubility in water: .. Other solubilities: soluble in dilute alkali,soluble in most common organic solvents
Formula Weight 167
Percent Purity 98%
Physical Form Low Melting Crystals
Chemical Name or Material 4-Bromobutyric acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-083-9
TSCA TSCA

RUO – Research Use Only