missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobutyl Acetate 97.0+%, TCI America™
$44.28 - $148.25
Chemical Identifiers
| CAS | 4753-59-7 |
|---|---|
| Molecular Formula | C6H11BrO2 |
| Molecular Weight (g/mol) | 195.056 |
| MDL Number | MFCD00000263 |
| InChI Key | UOABIRUEGSGTSA-UHFFFAOYSA-N |
| Synonym | 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide |
| PubChem CID | 78491 |
| IUPAC Name | 4-bromobutyl acetate |
| SMILES | CC(=O)OCCCCBr |
Chemical Identifiers
| 4753-59-7 | |
| 195.056 | |
| UOABIRUEGSGTSA-UHFFFAOYSA-N | |
| 78491 | |
| CC(=O)OCCCCBr |
| C6H11BrO2 | |
| MFCD00000263 | |
| 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
| 4-bromobutyl acetate |
Specifications
| 4753-59-7 | |
| 211°C | |
| MFCD00000263 | |
| 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
| CC(=O)OCCCCBr | |
| 195.056 | |
| 195.06 | |
| Liquid |
| Yellow | |
| C6H11BrO2 | |
| 5 g | |
| UOABIRUEGSGTSA-UHFFFAOYSA-N | |
| 4-bromobutyl acetate | |
| 78491 | |
| ≥97.0% (GC) | |
| 4-Bromobutyl Acetate |
Safety and Handling
TSCA : Yes