missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyl Acetate 97.0+%, TCI America™

Catalog No. B49585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49585G 5 g
B495825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49585G Supplier TCI America Supplier No. B49585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4753-59-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000263
InChI Key UOABIRUEGSGTSA-UHFFFAOYSA-N
Synonym 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide
PubChem CID 78491
IUPAC Name 4-bromobutyl acetate
SMILES CC(=O)OCCCCBr

Specifications

Color Yellow
Boiling Point 211°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromobutyl Acetate