missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobiphenyl, 98+%

Catalog No. AAA1263830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1263830 250 g
AAA1263809 10 g
AAA1263818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1263830 Supplier Thermo Scientific Chemicals Supplier No. A1263830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 92-66-0 | C12H9Br | 233.108 g/mol

4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobiphenyl is used as halogenated hydrocarbon. It is also used in used in the synthesis of 4-biphenylthiolate.

Solubility
Insoluble in water.

Notes
Sensitive to light. Store in cool dry place in tightly closed container. Avoid excessive heat and light. Store away from oxidizing agent.

Chemical Identifiers

CAS 92-66-0
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.108
MDL Number MFCD00000100
InChI Key PKJBWOWQJHHAHG-UHFFFAOYSA-N
Synonym 4-bromobiphenyl, p-bromobiphenyl, 4-bromo-1,1'-biphenyl, 4-bromodiphenyl, 1,1'-biphenyl, 4-bromo, 4-biphenyl bromide, p-bromodiphenyl, biphenyl, 4-bromo, 4-bromo-biphenyl, p-phenylbromobenzene
PubChem CID 7101
IUPAC Name 1-bromo-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)Br

Specifications

Melting Point 88°C to 92°C
Boiling Point 310°C
Flash Point 143°C (289°F)
Quantity 250 g
Beilstein 1907453
Solubility Information Insoluble in water.
Formula Weight 233.12
Percent Purity ≥98%
Chemical Name or Material 4-Bromobiphenyl
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-176-6
RTECSNumber DV1750100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only