missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7119298
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18415G 5 g
B184125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18415G Supplier TCI America Supplier No. B18415G
Only null left

Chemical Identifiers

CAS 26177-44-6
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012860
InChI Key BVYUYDBWQMSOKM-UHFFFAOYSA-N
Synonym 4-bromobenzylamine hydrochloride, 4-bromophenyl methanamine hydrochloride, 4-bromobenzylamine hcl, 4-bba x hcl, 4-bromo-benzylamine hydrochloride, p-bromobenzylamine hydrochloride, benzenemethanamine, 4-bromo-, hydrochloride, pubchem3751, acmc-1cfrz, 4-bromobenzylamine, hcl
PubChem CID 2724096
IUPAC Name hydrogen 1-(4-bromophenyl)methanamine chloride
SMILES [H+].[Cl-].NCC1=CC=C(Br)C=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 222.51
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzylamine Hydrochloride
EINECSNumber (3)-0316
TSCA No