missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine hydrochloride, 98%

Catalog No. AAA1097909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1097909 10 g
AAA1097918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1097909 Supplier Thermo Scientific Chemicals Supplier No. A1097909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26177-44-6 | C7H9BrClN | 222.51 g/mol

4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzylamine hydrochloride is used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine. Further, it is used to prepare 1-(4-Brombenzyl)-pyrrole by reacting with 2,5-dimethoxy-tetrahydro-furan.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 26177-44-6
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012860
InChI Key BVYUYDBWQMSOKM-UHFFFAOYSA-N
Synonym 4-bromobenzylamine hydrochloride, 4-bromophenyl methanamine hydrochloride, 4-bromobenzylamine hcl, 4-bba x hcl, 4-bromo-benzylamine hydrochloride, p-bromobenzylamine hydrochloride, benzenemethanamine, 4-bromo-, hydrochloride, pubchem3751, acmc-1cfrz, 4-bromobenzylamine, hcl
PubChem CID 2724096
IUPAC Name (4-bromophenyl)methanamine;hydrochloride
SMILES [H+].[Cl-].NCC1=CC=C(Br)C=C1

Specifications

Melting Point 272°C to 274°C
Quantity 10 g
Beilstein 3693458
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 222.52
Percent Purity 98%
Chemical Name or Material 4-Bromobenzylamine hydrochloride
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 247-503-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only