missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobenzyl)triphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. B52415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52415G 5 g
B524125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52415G Supplier TCI America Supplier No. B52415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51044-13-4
Molecular Formula C25H21Br2P
Molecular Weight (g/mol) 512.23
MDL Number MFCD00051922
InChI Key FQJYKXVQABPCRA-UHFFFAOYSA-M
Synonym 4-bromobenzyl triphenylphosphonium bromide, phosphonium, 4-bromophenyl methyl triphenyl-, bromide, 4-bromophenyl methyl triphenylphosphanium bromide, acmc-20apnl, 4-bromobenzyltriphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphonium-bromide, 4-bromobenzyl triphenyl-phosphonium bromide, 4-bromobenzyl triphenyl phosphonium bromide
PubChem CID 2724858
IUPAC Name [(4-bromophenyl)methyl]triphenylphosphanium bromide
SMILES [Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 275°C
Color White
Quantity 5 g
Formula Weight 512.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromobenzyl)triphenylphosphonium Bromide