missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobenzyl)triphenylphosphonium bromide, 98%

Catalog No. p-7022437
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522006 5 g
AAA1522014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522006 Supplier Thermo Scientific Chemicals Supplier No. A1522006

CAS: 51044-13-4 | C25H21Br2P | 512.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51044-13-4
Molecular Formula C25H21Br2P
Molecular Weight (g/mol) 512.23
MDL Number MFCD00051922
InChI Key FQJYKXVQABPCRA-UHFFFAOYSA-M
Synonym 4-bromobenzyl triphenylphosphonium bromide, phosphonium, 4-bromophenyl methyl triphenyl-, bromide, 4-bromophenyl methyl triphenylphosphanium bromide, acmc-20apnl, 4-bromobenzyltriphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphonium-bromide, 4-bromobenzyl triphenyl-phosphonium bromide, 4-bromobenzyl triphenyl phosphonium bromide
PubChem CID 2724858
IUPAC Name (4-bromophenyl)methyl-triphenylphosphanium;bromide
SMILES [Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 270°C to 273°C
Quantity 5 g
Beilstein 4173386
Sensitivity Hygroscopic
Formula Weight 512.24
Percent Purity 98%
Chemical Name or Material (4-Bromobenzyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only