missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl mercaptan, 98%

CAS: 19552-10-4 | C7H7BrS | 203.097 g/mol

Supplier:  Thermo Scientific Chemicals H2629906

 View more versions of this product

Catalog No. AAH2629906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromobenzyl mercaptan
26°C to 30°C
C7H7BrS
MFCD03701566
UN2811
4-bromobenzyl mercaptan, 4-bromophenyl methanethiol, 4-bromobenzylmercaptan, 4-bromophenyl methane-1-thiol, acmc-20ammu, pubchem6868, 4-bromo-a-toluenethiol, 4-bromo-, a-toluenethiol, 4-bromo-alpha-toluenethiol
C1=CC(=CC=C1CS)Br
203.097
203.1
19552-10-4
>110°C (230°F)
1.617
5 g
Air Sensitive
CUCKXDPCCYHFMQ-UHFFFAOYSA-N
(4-bromophenyl)methanethiol
271218
98%

Chemical Identifiers

19552-10-4
203.097
CUCKXDPCCYHFMQ-UHFFFAOYSA-N
271218
C1=CC(=CC=C1CS)Br
C7H7BrS
MFCD03701566
4-bromobenzyl mercaptan, 4-bromophenyl methanethiol, 4-bromobenzylmercaptan, 4-bromophenyl methane-1-thiol, acmc-20ammu, pubchem6868, 4-bromo-a-toluenethiol, 4-bromo-, a-toluenethiol, 4-bromo-alpha-toluenethiol
(4-bromophenyl)methanethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only