missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzotrifluoride, 99%

Catalog No. AC166670050
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166670050 5 mL
AC166670250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166670050 Supplier Thermo Scientific Chemicals Supplier No. 166670050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 402-43-7 | C7H4BrF3 | 225.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-43-7
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00000398
InChI Key XLQSXGGDTHANLN-UHFFFAOYSA-N
Synonym 4-bromobenzotrifluoride, 1-bromo-4-trifluoromethyl benzene, benzene, 1-bromo-4-trifluoromethyl, p-bromobenzotrifluoride, 4-bromotrifluorotoluene, 4-bromo-alpha,alpha,alpha-trifluorotoluene, p-bromotrifluoromethylbenzene, 4-trifluoromethyl bromobenzene, 4-bromo-a,a,a-trifluorotoluene, 4-trifluoromethylbromobenzene
PubChem CID 67872
IUPAC Name 1-bromo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 154.0°C to 155.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.4740
Linear Formula BrC6H4CF3
Quantity 5 mL
Specific Gravity 1.6
Formula Weight 225.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzotrifluoride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-943-6

RUO – Research Use Only